tert-Butyl-2-cyano-1H-pyrrol-1-ylcarbamate ≥98%

Supplier: Ambeed
Warning

A199211-5G A199211-25G A199211-10G A199211-1G
77671-762EA 27 USD
77671-762 77883-449 77883-447 77883-448
tert-Butyl-2-cyano-1H-pyrrol-1-ylcarbamate ≥98%
tert-Butyl-2-cyano-1H-pyrrol-1-ylcarbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₃N₃O₂
MDL Number: MFCD12068393
CAS Number: 937046-96-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR