H-Met-Pna ≥97%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
Synonyms:
L-Methionine p-nitroanilide, (S)-2-Amino-4-(methylthio)-N-(4-nitrophenyl)butanamide
A199011-250MG
A199011-100MG
A199011-1G
77611-912EA
32.96
USD
77611-912
77683-550
77890-203
H-Met-Pna ≥97%, Ambeed.Inc
H-Met-Pna
Precaución: For research use only.
Formula:
C₁₁H₁₅N₃O₃S MW: 269.319 g/mol |
MDL Number:
MFCD00063424 CAS Number: 6042-04-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...