5-Cyano-2-methoxybenzaldehyde ≥98%

Supplier: Ambeed
Warning

Synonyms: 3-Formyl-4-methoxybenzonitrile

A198785-1G A198785-5G
77683-548EA 24 USD
77683-548 77683-549
5-Cyano-2-methoxybenzaldehyde ≥98%
5-Cyano-2-methoxybenzaldehyde

Precaución: For research use only.

Formula: C₉H₇NO₂
MW: 161.16 g/mol
MDL Number: MFCD13195275
CAS Number: 21962-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR