Taurocholic acid sodium salt ≥98%

Supplier: Ambeed
Warning

Synonyms: Cholaic acid sodium salt, Taurocholic acid sodium, Sodium 2-((R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanamido)ethane-1-sulphonate (1:1), Sodium taurocholate

A198685-1G A198685-5G A198685-100G
77600-614EA 26.65 USD
77600-614 77600-616 77871-989
Taurocholic acid sodium salt ≥98%
Taurocholic acid sodium salt

Precaución: for Research Use Only

Formula: C₂₆H₄₄NNaO₇S
MDL Number: MFCD00013050
CAS Number: 145-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR