Fmoc-D-Ser(BSi)-OH 95+%
Supplier: Ambeed
|
Danger
|
Synonyms:
(R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-((tert-butyldimethylsilyl)oxy)propanoic acid, Fmoc-D-Ser(TBDMS)-OH
A197276-100MG
A197276-250MG
77901-975EA
9
USD
77901-975
77901-811
Fmoc-D-Ser(BSi)-OH 95+%
Fmoc-D-Ser(BSi)-OH
Precaución: for Research Use Only
Formula:
C₂₄H₃₁NO₅Si |
MDL Number:
MFCD08276730 CAS Number: 201210-25-5 UN: 2811 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...