tert-Butyl hexahydropyrrolo[3,4-b]pyrrole-1(2H)-carboxylate ≥97%, Ambeed.Inc

Supplier: Ambeed
Warning

A196992-100MG A196992-250MG
77683-540EA 121 USD
77683-540 77683-541
tert-Butyl hexahydropyrrolo[3,4-b]pyrrole-1(2H)-carboxylate ≥97%, Ambeed.Inc
tert-Butyl hexahydropyrrolo[3,4-b]pyrrole-1(2H)-carboxylate

Precaución: For research use only.

Formula: C₁₁H₂₀N₂O₂
MW: 212.29 g/mol
MDL Number: MFCD06739059
CAS Number: 185693-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR