1,2,5,6-Tetrahydropyridine-3-carboxylic acid hydrochloride ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A196707-10MG A196707-5MG A196707-100MG A196707-250MG A196707-50MG
77683-538EA 51 USD
77683-538 77683-539 77885-452 77885-453 77885-454
1,2,5,6-Tetrahydropyridine-3-carboxylic acid hydrochloride ≥95%, Ambeed.Inc
1,2,5,6-Tetrahydropyridine-3-carboxylic acid hydrochloride

Precaución: For research use only.

Formula: C₆H₁₀ClNO₂
MDL Number: MFCD00055191
CAS Number: 6027-91-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR