(R)-4-Hydroxy-2,6-bis(2,4,6-tri-tert-butylphenyl)dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide ≥95%

Supplier: Ambeed
Warning

A1963833-50MG A1963833-100MG
77593-272EA 203.98 USD
77593-272 77625-972
(R)-4-Hydroxy-2,6-bis(2,4,6-tri-tert-butylphenyl)dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide ≥95%
(R)-4-Hydroxy-2,6-bis(2,4,6-tri-tert-butylphenyl)dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide

Precaución: for Research Use Only

Formula: C₅₆H₆₉O₄P
MW: 837.12 g/mol
CAS Number: 1821241-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR