1-Butyl-3-chloro-4-(4-phenylpiperidin-1-yl)pyridin-2(1H)-one 98+%

Supplier: Ambeed
Warning

Synonyms: JNJ-40411813

A196362-1MG A196362-5MG A196362-10MG
77615-662EA 9.27 USD
77615-662 77615-664 77615-660
1-Butyl-3-chloro-4-(4-phenylpiperidin-1-yl)pyridin-2(1H)-one 98+%
1-Butyl-3-chloro-4-(4-phenylpiperidin-1-yl)pyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₂₀H₂₅ClN₂O
MW: 344.88 g/mol
MDL Number: MFCD22572363
CAS Number: 1127498-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR