Potassium (E)-(3,3-dimethylbut-1-en-1-yl)trifluoroborate ≥95%

Supplier: Ambeed
Danger

A1963386-1G A1963386-250MG A1963386-5G
77877-563EA 119 USD
77877-563 77877-564 77877-565
Potassium (E)-(3,3-dimethylbut-1-en-1-yl)trifluoroborate ≥95%
Potassium (E)-(3,3-dimethylbut-1-en-1-yl)trifluoroborate

Precaución: for Research Use Only

Formula: C₆H₁₁BF₃K
MW: 190.06 g/mol
MDL Number: MFCD32662885
CAS Number: 1273440-63-3
UN: 1759
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR