N-(3-Aminopropyl)-3-(3,4-dihydroxyphenyl)-N-(4-((3-(3-(3,4-dihydroxyphenyl)propanamido)propyl)amino)butyl)propanamide ≥97%

Supplier: Ambeed
Warning

A196244-10MG A196244-5MG
77748-244EA 118 USD
77748-244 77748-245
N-(3-Aminopropyl)-3-(3,4-dihydroxyphenyl)-N-(4-((3-(3-(3,4-dihydroxyphenyl)propanamido)propyl)amino)butyl)propanamide ≥97%
N-(3-Aminopropyl)-3-(3,4-dihydroxyphenyl)-N-(4-((3-(3-(3,4-dihydroxyphenyl)propanamido)propyl)amino)butyl)propanamide

Precaución: for Research Use Only

Formula: C₂₈H₄₂N₄O₆
MDL Number: MFCD29904539
CAS Number: 164991-67-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR