(R)-N-(2-(1H-Indol-3-yl)-1-(4-(4-methoxybenzyl)-5-phenethyl-4H-1,2,4-triazol-3-yl)ethyl)-2-aminoacetamide hydrochloride ≥98%

Supplier: Ambeed
Warning

Synonyms: JMV 2959 hydrochloride

A1961316-10MG A1961316-1MG A1961316-5MG
77598-274EA 360.8 USD
77598-274 77598-276 77598-278
(R)-N-(2-(1H-Indol-3-yl)-1-(4-(4-methoxybenzyl)-5-phenethyl-4H-1,2,4-triazol-3-yl)ethyl)-2-aminoacetamide hydrochloride ≥98%
(R)-N-(2-(1H-Indol-3-yl)-1-(4-(4-methoxybenzyl)-5-phenethyl-4H-1,2,4-triazol-3-yl)ethyl)-2-aminoacetamide hydrochloride

Precaución: for Research Use Only

Formula: C₃₀H₃₃ClN₆O₂
MDL Number: MFCD31715444
CAS Number: 2448414-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR