Potassium (E)-(2-cyclohexylvinyl)trifluoroborate ≥95%
Supplier: Ambeed
|
Danger
|
A1961204-250MG
A1961204-5G
A1961204-1G
77877-537EA
58
USD
77877-537
77877-538
77877-536
Potassium (E)-(2-cyclohexylvinyl)trifluoroborate ≥95%
Potassium (E)-(2-cyclohexylvinyl)trifluoroborate
Precaución: for Research Use Only
Formula:
C₈H₁₃BF₃K |
CAS Number:
1005201-61-5 UN: 3261 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...