Potassium trifluoro(2-methoxyethyl)borate ≥98%

Supplier: Ambeed
Warning

A195991-1G A195991-250MG
77892-262EA 576 USD
77892-262 77892-263
Potassium trifluoro(2-methoxyethyl)borate ≥98%
Potassium trifluoro(2-methoxyethyl)borate

Precaución: for Research Use Only

Formula: C₃H₇BF₃KO
MDL Number: MFCD18861717
CAS Number: 1408168-69-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR