Gentiopicroside ≥99%

Supplier: Ambeed
Warning

Synonyms: (5R,6S)-6-(((2S,3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-5-vinyl-5,6-dihydro-1H,3H-pyrano[3,4-c]pyran-1-one, Gentiopicrin

A195939-10MG A195939-25MG A195939-5MG A195939-1MG
77593-264EA 9.23 USD
77593-264 77593-266 77593-268 77685-488
Gentiopicroside ≥99%
Gentiopicroside

Precaución: for Research Use Only

Formula: C₁₆H₂₀O₉
MW: 356.33 g/mol
MDL Number: MFCD00075700
CAS Number: 20831-76-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR