4-Nitro-1H-pyrazole-3-carboxamide ≥95%

Supplier: Ambeed
Warning

A195722-1G A195722-250MG
77881-440EA 97 USD
77881-440 77881-441
4-Nitro-1H-pyrazole-3-carboxamide ≥95%
4-Nitro-1H-pyrazole-3-carboxamide

Precaución: for Research Use Only

Formula: C₄H₄N₄O₃
MDL Number: MFCD00619107
CAS Number: 65190-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR