3-Chloropyridazin-4-amine ≥98%

Supplier: Ambeed
Warning

A195619-100MG A195619-250MG
77871-976EA 201 USD
77871-976 77871-977
3-Chloropyridazin-4-amine ≥98%
3-Chloropyridazin-4-amine

Precaución: for Research Use Only

Formula: C4H4ClN3
MW: 129.55 g/mol
MDL Number: MFCD09834368
CAS Number: 55928-83-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR