2-[[[4-(3-Methoxypropoxy)-3-methylpyridine-2-yl ]methyl]thio]-1H-benzimidazole ≥97%

Supplier: Ambeed
Warning

Synonyms: Rabeprazole sulphide, 2-[[4-(3-Methoxypropoxy)-3-methyl-2-pyridyl]methylthio]benzimidazole

A195582-100G A195582-25G
77600-590EA 105.58 USD
77600-590 77671-734
2-[[[4-(3-Methoxypropoxy)-3-methylpyridine-2-yl ]methyl]thio]-1H-benzimidazole ≥97%
2-[[[4-(3-Methoxypropoxy)-3-methylpyridine-2-yl ]methyl]thio]-1H-benzimidazole

Precaución: for Research Use Only

Formula: C₁₈H₂₁N₃O₂S
MW: 343.4432 g/mol
MDL Number: MFCD08063845
CAS Number: 117977-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR