(2-Cyclopropylpyrimidin-5-yl)boronic acid ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A195223-100MG A195223-250MG
77683-530EA 36 USD
77683-530 77683-531
(2-Cyclopropylpyrimidin-5-yl)boronic acid ≥98%, Ambeed.Inc
(2-Cyclopropylpyrimidin-5-yl)boronic acid

Precaución: For research use only.

Formula: C₇H₉BN₂O₂
MW: 163.97 g/mol
MDL Number: MFCD12032614
CAS Number: 893567-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR