4-Bromo-2-cyclopropoxypyridine ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A194984-1G A194984-5G A194984-100MG A194984-250MG
77683-526EA 73 USD
77683-526 77890-194 77890-192 77890-193
4-Bromo-2-cyclopropoxypyridine ≥95%, Ambeed.Inc
4-Bromo-2-cyclopropoxypyridine

Precaución: For research use only.

Formula: C₈H₈BrNO
MDL Number: MFCD14702588
CAS Number: 1209460-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR