2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide ≥95%

Supplier: Ambeed
Warning

A1949303-5G A1949303-1G A1949303-250MG
77877-527EA 244 USD
77877-527 77877-525 77877-526
2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide ≥95%
2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

Precaución: for Research Use Only

Formula: C₁₃H₁₇BClNO₃
MDL Number: MFCD18383425
CAS Number: 1256958-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR