2,3-Dibromo-1,4-dimethoxybenzene ≥98%

Supplier: Ambeed
Warning

A194860-5G A194860-1G A194860-100MG A194860-250MG
77593-256EA 255.23 USD
77593-256 77611-870 77683-524 77683-525
2,3-Dibromo-1,4-dimethoxybenzene ≥98%
2,3-Dibromo-1,4-dimethoxybenzene

Precaución: For research use only.

Formula: C₈H₈Br₂O₂
MW: 295.96 g/mol
MDL Number: MFCD11519213
CAS Number: 5030-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR