1,2-Dimyristoyl-sn-glycero-3-phospho-rac-(1-glycerol) sodium salt ≥98%

Supplier: Ambeed
Warning

Synonyms: 1,2-Dimyristoyl-sn-glycero-3-phospho-rac-(1-glycerol) Sodium Salt, DMPG Sodium Salt, Sodium (R)-2,3-bis(tetradecanoyloxy)propyl (2,3-dihydroxypropyl) phosphate, Dimyristoylphosphatidylglycerol Sodium Salt, Sodium (R)-2,3-Bis(tetradecanoyloxy)propyl 2,3-Dihydroxypropyl Phosphate

A194556-1G A194556-250MG
77611-860EA 109.18 USD
77611-860 77685-476
1,2-Dimyristoyl-sn-glycero-3-phospho-rac-(1-glycerol) sodium salt ≥98%
1,2-Dimyristoyl-sn-glycero-3-phospho-rac-(1-glycerol) sodium salt

Precaución: for Research Use Only

Formula: C₃₄H₆₇O₁₀P
MW: 688.86 g/mol
MDL Number: MFCD00056649
CAS Number: 200880-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR