Dizocilpine maleate ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: (5S,10R)-5-Methyl-10,11-dihydro-5H-5,10-epiminodibenzo[a,d][7]annulene maleate, 10,11-dihydro-5S-methyl-5H-dibenzo[a,d]cyclohepten-5,10-imine, (2Z)-2-butenedioate,Dizocilpine, (+)-MK 801 hydrogen maleate

A194394-1MG A194394-50MG A194394-100MG A194394-250MG
77611-858EA 11.33 USD
77611-858 77683-522 77892-255 77892-256
Dizocilpine maleate ≥98%, Ambeed.Inc
Dizocilpine maleate

Precaución: For research use only.

Formula: C₁₆H₁₅N·C₄H₄O₄
MW: 337.37 g/mol
MDL Number: MFCD00082465
CAS Number: 77086-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR