2-(Methoxymethyl)-morpholine hydrochloride ≥97%

Supplier: Ambeed
Warning

Synonyms: 2-(Methoxymethyl)morpholine hydrochloride

A194278-250MG A194278-100MG A194278-1G
77593-248EA 59.45 USD
77593-248 77683-520 77683-521
2-(Methoxymethyl)-morpholine hydrochloride ≥97%
2-(Methoxymethyl)-morpholine hydrochloride

Precaución: For research use only.

Formula: C₆H₁₃NO₂·ClH
MW: 167.634 g/mol
MDL Number: MFCD16294690
CAS Number: 144053-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR