4-((2R,3S,4S,5R)-3-(3,4-Difluoro-2-methoxyphenyl)-4,5-dimethyl-5-(trifluoromethyl)tetrahydrofuran-2-carboxamido)picolinamide ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Suzetrigine
A1942314-1MG
A1942314-5MG
77625-952EA
448.05
USD
77625-952
77625-954
4-((2R,3S,4S,5R)-3-(3,4-Difluoro-2-methoxyphenyl)-4,5-dimethyl-5-(trifluoromethyl)tetrahydrofuran-2-carboxamido)picolinamide ≥98%
4-((2R,3S,4S,5R)-3-(3,4-Difluoro-2-methoxyphenyl)-4,5-dimethyl-5-(trifluoromethyl)tetrahydrofuran-2-carboxamido)picolinamide
Precaución: for Research Use Only
Formula:
C₂₁H₂₀F₅N₃O₄ MW: 473.39 g/mol |
CAS Number:
2649467-58-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...