1H-Pyrazolo[3,4-d]pyrimidin-6-amine ≥95%

Supplier: Ambeed
Warning

A194182-100MG A194182-250MG
77890-188EA 88 USD
77890-188 77890-189
1H-Pyrazolo[3,4-d]pyrimidin-6-amine ≥95%
1H-Pyrazolo[3,4-d]pyrimidin-6-amine

Precaución: for Research Use Only

Formula: C₅H₅N₅
MDL Number: MFCD11559009
CAS Number: 287177-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR