Binimetinib ≥98%

Supplier: Ambeed
Warning

A194061-5MG A194061-10MG A194061-1MG A194061-1G A194061-250MG
77611-854EA 14.42 USD
77611-854 77685-472 77685-473 77900-572 77901-114
Binimetinib ≥98%
Binimetinib

Precaución: for Research Use Only

Formula: C₁₇H₁₅BrF₂N₄O₃
MDL Number: MFCD22124525
CAS Number: 606143-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR