5-Fluoro-2-(trifluoromethyl)pyridine ≥98%

Supplier: Ambeed
Danger

A194014-250MG A194014-1G
77598-264EA 18.45 USD
77598-264 77877-493
5-Fluoro-2-(trifluoromethyl)pyridine ≥98%
5-Fluoro-2-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C6H3F4N
MDL Number: MFCD09864698
CAS Number: 936841-73-5
UN: 1993
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR