4-Chloro-2,3-difluoroaniline ≥98%

Supplier: Ambeed
Warning

A193953-1G A193953-250MG
77671-718EA 12 USD
77671-718 77671-720
4-Chloro-2,3-difluoroaniline ≥98%
4-Chloro-2,3-difluoroaniline

Precaución: for Research Use Only

Formula: C₆H₄ClF₂N
MW: 163.5500 g/mol
MDL Number: MFCD08459284
CAS Number: 878285-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR