((2R,3R)-3-((S)-1-((3R,4S,5S)-4-((S)-2-((S)-2-((4-((tert-Butoxycarbonyl)(methyl)amino)butyl)(methyl)amino)-3-methylbutanamido)-N,3-dimethylbutanamido)-3-methoxy-5-methylheptanoyl)pyrrolidin-2-yl)-3-methoxy-2-methylpropanoyl)-L-phenylalanine ≥95%

Supplier: Ambeed
Warning

Synonyms: Auristatin23

A1938486-25MG A1938486-100MG A1938486-50MG
77598-256EA 331.08 USD
77598-256 77625-944 77625-946
((2R,3R)-3-((S)-1-((3R,4S,5S)-4-((S)-2-((S)-2-((4-((tert-Butoxycarbonyl)(methyl)amino)butyl)(methyl)amino)-3-methylbutanamido)-N,3-dimethylbutanamido)-3-methoxy-5-methylheptanoyl)pyrrolidin-2-yl)-3-methoxy-2-methylpropanoyl)-L-phenylalanine ≥95%
((2R,3R)-3-((S)-1-((3R,4S,5S)-4-((S)-2-((S)-2-((4-((tert-Butoxycarbonyl)(methyl)amino)butyl)(methyl)amino)-3-methylbutanamido)-N,3-dimethylbutanamido)-3-methoxy-5-methylheptanoyl)pyrrolidin-2-yl)-3-methoxy-2-methylpropanoyl)-L-phenylalanine

Precaución: for Research Use Only

Formula: C₄₉H₈₄N₆O₁₀
MW: 917.23 g/mol
CAS Number: 2375388-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR