2-(8-Ethynyl-7-fluoro-3-(methoxymethoxy)naphthalen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95%

Supplier: Ambeed
Warning

A1938440-1G A1938440-250MG A1938440-100MG
77877-487EA 712 USD
77877-487 77877-488 77877-486
2-(8-Ethynyl-7-fluoro-3-(methoxymethoxy)naphthalen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95%
2-(8-Ethynyl-7-fluoro-3-(methoxymethoxy)naphthalen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₂₀H₂₂BFO₄
CAS Number: 2858674-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR