Oxfendazole ≥99%

Supplier: Ambeed
Warning

Synonyms: Methyl (6-(phenylsulphinyl)-1H-benzo[d]imidazol-2-yl)carbamate, Methyl [5-(Phenylsulphinyl)benzimidazol-2-yl]carbamate, [5-(Phenylsulphinyl)benzimidazol-2-yl]carbamic acid Methyl Ester

A193817-5G A193817-100G
77600-576EA 12.3 USD
77600-576 77667-618
Oxfendazole ≥99%
Oxfendazole

Precaución: for Research Use Only

Formula: C₁₅H₁₃N₃O₃S
MW: 315.35 g/mol
MDL Number: MFCD00801063
CAS Number: 53716-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR