2-((tert-Butyldiphenylsilyl)oxy)acetic acid ≥97%

Supplier: Ambeed
Warning

A193574-100MG A193574-1G A193574-250MG
77885-435EA 53 USD
77885-435 77885-436 77885-437
2-((tert-Butyldiphenylsilyl)oxy)acetic acid ≥97%
2-((tert-Butyldiphenylsilyl)oxy)acetic acid

Precaución: for Research Use Only

Formula: C₁₈H₂₂O₃Si
MDL Number: MFCD20039760
CAS Number: 76271-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR