2-(Dimethylamino)ethyl acrylate ≥98% stabilized

Supplier: Ambeed
Danger


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A193390-100G A193390-25G
77879-226EA 17 USD
77879-226 77879-227
2-(Dimethylamino)ethyl acrylate ≥98% stabilized
2-(Dimethylamino)ethyl acrylate

Precaución: for Research Use Only

Formula: C₇H₁₃NO₂
MDL Number: MFCD00038233
CAS Number: 2439-35-2
UN: 3302
ADR: 6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR