1-(tert-Butyl) 4-methyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1,4-dicarboxylate ≥95%
Supplier: Ambeed
|
Warning
|
A1933519-100MG
A1933519-1G
A1933519-250MG
77625-934EA
167.89
USD
77625-934
77625-936
77625-938
1-(tert-Butyl) 4-methyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1,4-dicarboxylate ≥95%
1-(tert-Butyl) 4-methyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1,4-dicarboxylate
Precaución: for Research Use Only
Formula:
C₂₁H₂₈BNO₆ MW: 401.26 g/mol |
MDL Number:
MFCD16036141 CAS Number: 1256359-97-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...