N-(2-Chloro-4-(trifluoromethyl)phenyl)-2-(2-(3,6-dihydro-2H-pyran-4-yl)-5-ethyl-6-(4-(5-hydroxy-6-methylpyrimidine-4-carbonyl)piperazin-1-yl)-7-oxo-[1,2,4]triazolo[1,5-a]pyrimidin-4(7H)-yl)acetamide ≥98%

Supplier: Ambeed
Warning

A1929330-10MG A1929330-1MG A1929330-5MG A1929330-100MG A1929330-25MG A1929330-50MG
77598-248EA 168.1 USD
77598-248 77598-250 77598-252 77877-441 77877-442 77877-443
N-(2-Chloro-4-(trifluoromethyl)phenyl)-2-(2-(3,6-dihydro-2H-pyran-4-yl)-5-ethyl-6-(4-(5-hydroxy-6-methylpyrimidine-4-carbonyl)piperazin-1-yl)-7-oxo-[1,2,4]triazolo[1,5-a]pyrimidin-4(7H)-yl)acetamide ≥98%
N-(2-Chloro-4-(trifluoromethyl)phenyl)-2-(2-(3,6-dihydro-2H-pyran-4-yl)-5-ethyl-6-(4-(5-hydroxy-6-methylpyrimidine-4-carbonyl)piperazin-1-yl)-7-oxo-[1,2,4]triazolo[1,5-a]pyrimidin-4(7H)-yl)acetamide

Precaución: for Research Use Only

Formula: C₃₁H₃₁ClF₃N₉O₅
CAS Number: 2869954-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR