Quinine sulfate monohydrate ≥95%

Supplier: Ambeed
Warning

Synonyms: Quinine sulphate hydrate, (R)-(6-methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methanol sulphate hydrate(2:1:4)

A192815-25G A192815-5G
77685-467EA 34 USD
77685-467 77685-468
Quinine sulfate monohydrate ≥95%
Quinine sulfate monohydrate

Precaución: for Research Use Only

Formula: C₄₀H₅₈N₄O₁₂S
MW: 818.97 g/mol
MDL Number: MFCD29075629
CAS Number: 6119-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR