(1-Phenyl-1H-pyrazol-3-yl)boronic acid ≥95%

Supplier: Ambeed
Warning

A1926959-100MG A1926959-1G A1926959-250MG
77877-431EA 37 USD
77877-431 77877-432 77877-433
(1-Phenyl-1H-pyrazol-3-yl)boronic acid ≥95%
(1-Phenyl-1H-pyrazol-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₉BN₂O₂
CAS Number: 1802503-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR