(3S,4S,5R)-4-((2R,3R)-2,3-Dihydroxy-3-((2S,3R,5R,9R,10R,13R,17S)-2,3,14-trihydroxy-10,13-dimethyl-6-oxo-2,3,4,5,6,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl)butyl)-5-hydroxy-3-methyltetrahydro-2H-pyran-2-one ≥97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Cyasterone
A192479-10MG
A192479-5MG
A192479-1MG
77593-234EA
43.05
USD
77593-234
77593-236
77625-926
(3S,4S,5R)-4-((2R,3R)-2,3-Dihydroxy-3-((2S,3R,5R,9R,10R,13R,17S)-2,3,14-trihydroxy-10,13-dimethyl-6-oxo-2,3,4,5,6,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl)butyl)-5-hydroxy-3-methyltetrahydro-2H-pyran-2-one ≥97%
(3S,4S,5R)-4-((2R,3R)-2,3-Dihydroxy-3-((2S,3R,5R,9R,10R,13R,17S)-2,3,14-trihydroxy-10,13-dimethyl-6-oxo-2,3,4,5,6,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl)butyl)-5-hydroxy-3-methyltetrahydro-2H-pyran-2-one
Precaución: for Research Use Only
Formula:
C₂₉H₄₄O₉ MW: 536.65 g/mol |
MDL Number:
MFCD01683622 CAS Number: 17086-76-9 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...