(4-fluoro-3-(morpholinosulfonyl)phenyl)boronic acid 98+%

Supplier: Ambeed
Warning

A192334-1G A192334-250MG A192334-50MG
77593-232EA 999.38 USD
77593-232 77892-240 77892-241
(4-fluoro-3-(morpholinosulfonyl)phenyl)boronic acid 98+%
(4-fluoro-3-(morpholinosulfonyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃BFNO₅S
MDL Number: MFCD28805611
CAS Number: 1704121-53-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR