(4-Bromo-2-fluoro-5-methylphenyl)boronic acid ≥97%

Supplier: Ambeed
Warning

A192166-100MG A192166-250MG
77883-411EA 48 USD
77883-411 77883-412
(4-Bromo-2-fluoro-5-methylphenyl)boronic acid ≥97%
(4-Bromo-2-fluoro-5-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₇BBrFO₂
MDL Number: MFCD20441808
CAS Number: 677777-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR