(2S,3R,4S,5S,6R)-2-(((2R,3R,4S,5S,6R)-3,5-Dihydroxy-2-(((3S,4R,4aR,6aR,6bS,8R,8aS,12aR,12bS,14aR,14bS)-8-hydroxy-4-(hydroxymethyl)-4,6a,6b,11,11,14b-hexamethyl-1,2,3,4,4a,5,6,6a,6b,7,8,9,10,11,12,12a,14a,14b-octadecahydro-12b,8a-(epoxymethano)picen-3-yl)oxy)-6-methyltetrahydro-2H-pyran-4-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol 98+%

Supplier: Ambeed
Warning

A191999-100MG A191999-50MG
77900-632EA 193 USD
77900-632 77900-509
(2S,3R,4S,5S,6R)-2-(((2R,3R,4S,5S,6R)-3,5-Dihydroxy-2-(((3S,4R,4aR,6aR,6bS,8R,8aS,12aR,12bS,14aR,14bS)-8-hydroxy-4-(hydroxymethyl)-4,6a,6b,11,11,14b-hexamethyl-1,2,3,4,4a,5,6,6a,6b,7,8,9,10,11,12,12a,14a,14b-octadecahydro-12b,8a-(epoxymethano)picen-3-yl)oxy)-6-methyltetrahydro-2H-pyran-4-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol 98+%
(2S,3R,4S,5S,6R)-2-(((2R,3R,4S,5S,6R)-3,5-Dihydroxy-2-(((3S,4R,4aR,6aR,6bS,8R,8aS,12aR,12bS,14aR,14bS)-8-hydroxy-4-(hydroxymethyl)-4,6a,6b,11,11,14b-hexamethyl-1,2,3,4,4a,5,6,6a,6b,7,8,9,10,11,12,12a,14a,14b-octadecahydro-12b,8a-(epoxymethano)picen-3-yl)

Precaución: for Research Use Only

Formula: C₄₂H₆₈O₁₃
MDL Number: MFCD09028095
CAS Number: 20874-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR