Acetylthiocholine chloride ≥98%

Supplier: Ambeed
Danger

A191863-1G A191863-5G
77593-224EA 43.05 USD
77593-224 77877-426
Acetylthiocholine chloride ≥98%
Acetylthiocholine chloride

Precaución: for Research Use Only

Formula: H₃CC(S)O(CH₂)₂N(CH₃)₃Cl
MDL Number: MFCD00038727
CAS Number: 6050-81-3
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR