LY-2940680 98+%

Supplier: Ambeed
Warning

Synonyms: 4-Fluoro-N-methyl-N-(1-(4-(1-methyl-1H-pyrazol-5-yl)phthalazin-1-yl)piperidin-4-yl)-2-(trifluoromethyl)benzamide, Taladegib

A191567-10MG A191567-1MG A191567-5MG
77685-460EA 37 USD
77685-460 77685-461 77685-462
LY-2940680 98+%
LY-2940680

Precaución: for Research Use Only

Formula: C₂₆H₂₄F₄N₆O
MW: 512.5 g/mol
MDL Number: MFCD21609264
CAS Number: 1258861-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR