(R)-tert-Butyl 2-(methoxymethyl)piperazine-1-carboxylate ≥97%

Supplier: Ambeed
Warning

A191536-100MG A191536-250MG
77593-220EA 50.23 USD
77593-220 77671-704
(R)-tert-Butyl 2-(methoxymethyl)piperazine-1-carboxylate ≥97%
(R)-tert-Butyl 2-(methoxymethyl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₃
MW: 230.3 g/mol
MDL Number: MFCD11112288
CAS Number: 1023301-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR