N1,N2-Dibenzylethane-1,2-diamine bis((6R,7R)-3-(acetoxymethyl)-8-oxo-7-(2-(pyridin-4-ylthio)acetamido)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate) ≥97%

Supplier: Ambeed
Danger

Synonyms: Cephapirin Benzathine

A191109-10MG A191109-1MG A191109-5MG
77615-626EA 8.24 USD
77615-626 77748-224 77748-225
N1,N2-Dibenzylethane-1,2-diamine bis((6R,7R)-3-(acetoxymethyl)-8-oxo-7-(2-(pyridin-4-ylthio)acetamido)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate) ≥97%
N1,N2-Dibenzylethane-1,2-diamine bis((6R,7R)-3-(acetoxymethyl)-8-oxo-7-(2-(pyridin-4-ylthio)acetamido)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate)

Precaución: for Research Use Only

Formula: C₅₀H₅₄N₈O₁₂S₄
MDL Number: MFCD09029421
CAS Number: 97468-37-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR