Dichloro[1,1'-bis(diisopropylphosphino)ferrocene]palladium(II) ≥98%

Supplier: Ambeed
Danger

A191042-100MG A191042-1G A191042-250MG
77685-456EA 11 USD
77685-456 77685-457 77901-214
Dichloro[1,1'-bis(diisopropylphosphino)ferrocene]palladium(II) ≥98%
Dichloro[1,1'-bis(diisopropylphosphino)ferrocene]palladium(II)

Precaución: for Research Use Only

Formula: C₂₂H₃₀Cl₂FeP₂Pd
MDL Number: MFCD11656082
CAS Number: 215788-65-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR