4-(1-Pyrrolidinyl)-1-butylamine ≥98%

Supplier: Ambeed
Danger

A191030-5G A191030-1G
77625-916EA 145.85 USD
77625-916 77877-423
4-(1-Pyrrolidinyl)-1-butylamine ≥98%
4-(1-Pyrrolidinyl)-1-butylamine

Precaución: for Research Use Only

Formula: C₈H₁₈N₂
MDL Number: MFCD00192591
CAS Number: 24715-90-0
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR