Vincamine ≥98%

Supplier: Ambeed
Warning

Synonyms: Vincamine base, Methyl (41S,12S,13aS)-13a-ethyl-12-hydroxy-2,3,41,5,6,12,13,13a-octahydro-1H-indolo[3,2,1-de]pyrido[3,2,1-ij][1,5]naphthyridine-12-carboxylate

A190417-1G A190417-25G A190417-5G
77600-552EA 44.08 USD
77600-552 77667-586 77667-588
Vincamine ≥98%
Vincamine

Precaución: for Research Use Only

Formula: C₂₁H₂₆N₂O₃
MW: 354.45 g/mol
MDL Number: MFCD00078054
CAS Number: 1617-90-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR